Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGATGCTGGAGAACTACAGGAAC[A/G]TGGTCTCCCTGGGTAAGGCAAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF554 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF554 - zinc finger protein 554 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102651.1 | 1406 | Missense Mutation | ATG,GTG | M,V 81 | NP_001096121.1 | |
XM_006722635.3 | 1406 | UTR 5 | XP_006722698.1 | |||
XM_011527660.2 | 1406 | Missense Mutation | ATG,GTG | M,V 30 | XP_011525962.1 | |
XM_011527662.2 | 1406 | Missense Mutation | ATG,GTG | M,V 30 | XP_011525964.1 | |
XM_011527664.2 | 1406 | UTR 5 | XP_011525966.1 | |||
XM_011527667.2 | 1406 | Missense Mutation | ATG,GTG | M,V 81 | XP_011525969.1 | |
XM_017026234.1 | 1406 | Missense Mutation | ATG,GTG | M,V 81 | XP_016881723.1 | |
XM_017026235.1 | 1406 | UTR 5 | XP_016881724.1 | |||
XM_017026236.1 | 1406 | Intron | XP_016881725.1 |