Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTTCTGGTTCCAGCTGAGCCA[C/T]ATCTGGTGCAGCTTCTCCTGGCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602702 | ||||||||||||||||||||
Literature Links: |
PLIN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLIN3 - perilipin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164189.1 | 1077 | Missense Mutation | ATA,ATG | I,M 300 | NP_001157661.1 | |
NM_001164194.1 | 1077 | Missense Mutation | ATA,ATG | I,M 288 | NP_001157666.1 | |
NM_005817.4 | 1077 | Missense Mutation | ATA,ATG | I,M 300 | NP_005808.3 |