Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGTGCTCCCGGCTAGCCGCGCCT[A/G]CGTCCATGGCACCCTGCCAGGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608066 | ||||||||||||||||||||
Literature Links: |
SAFB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAFB2 - scaffold attachment factor B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014649.2 | 2165 | Missense Mutation | GCA,GTA | A,V 868 | NP_055464.1 | |
XM_011528449.2 | 2165 | Missense Mutation | GCA,GTA | A,V 868 | XP_011526751.1 | |
XM_011528450.2 | 2165 | Missense Mutation | GCA,GTA | A,V 414 | XP_011526752.1 | |
XM_017027520.1 | 2165 | Missense Mutation | GCA,GTA | A,V 414 | XP_016883009.1 | |
XM_017027521.1 | 2165 | Missense Mutation | GCA,GTA | A,V 414 | XP_016883010.1 | |
XM_017027522.1 | 2165 | Missense Mutation | GCA,GTA | A,V 414 | XP_016883011.1 |