Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTGCAGGCCATCCACGGCTTC[A/C]GAGAGACAGAGAAGTCGCGCTGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613305 MIM: 601119 MIM: 189968 MIM: 602921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALKBH7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALKBH7 - alkB homolog 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032306.3 | 184 | Silent Mutation | AGA,CGA | R,R 74 | NP_115682.1 | |
XM_005259658.4 | 184 | Silent Mutation | AGA,CGA | R,R 48 | XP_005259715.1 | |
XM_017027355.1 | 184 | Silent Mutation | AGA,CGA | R,R 74 | XP_016882844.1 |
CLPP - caseinolytic mitochondrial matrix peptidase proteolytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GTF2F1 - general transcription factor IIF subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSPN - persephin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |