Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGAGCAGCCCAGCCAACCTCA[A/G]TGCCTCCGTCGCCACAGTCCAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 130130 MIM: 177020 | ||||||||||||||||||||
Literature Links: |
ELANE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELANE - elastase, neutrophil expressed | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRTN3 - proteinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002777.3 | 436 | Missense Mutation | AAT,AGT | N,S 131 | NP_002768.3 | |
XM_011528136.1 | 436 | Missense Mutation | AAT,AGT | N,S 131 | XP_011526438.1 |