Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGAACCTTCCGCGGAACCCCA[C/T]CCCCTCTCCTTGCTGACCAGCTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606651 MIM: 611011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIN3B - glutamate ionotropic receptor NMDA type subunit 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138690.2 | 2342 | Intron | NP_619635.1 | |||
XM_017026243.1 | 2342 | Intron | XP_016881732.1 |
LOC105372235 - uncharacterized LOC105372235 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM259 - transmembrane protein 259 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033026.1 | 2342 | UTR 3 | NP_001028198.1 | |||
NM_033420.3 | 2342 | UTR 3 | NP_219488.1 | |||
XM_005259675.2 | 2342 | UTR 3 | XP_005259732.1 | |||
XM_005259676.2 | 2342 | UTR 3 | XP_005259733.1 | |||
XM_005259677.2 | 2342 | UTR 3 | XP_005259734.1 | |||
XM_005259678.2 | 2342 | UTR 3 | XP_005259735.1 | |||
XM_017027457.1 | 2342 | UTR 3 | XP_016882946.1 |