Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTCATCGTGGCTCGTGGAACA[A/G]TGTCGGCAGCTGGGTTTGAAGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606942 MIM: 604800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPE - coatomer protein complex subunit epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007263.3 | 115 | Intron | NP_009194.2 | |||
NM_199442.1 | 115 | Intron | NP_955474.1 | |||
NM_199444.1 | 115 | Intron | NP_955476.1 | |||
XM_011527655.1 | 115 | Intron | XP_011525957.1 | |||
XM_011527656.1 | 115 | Intron | XP_011525958.1 |
DDX49 - DEAD-box helicase 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019070.4 | 115 | Silent Mutation | CAA,CAG | Q,Q 16 | NP_061943.2 | |
XM_011528083.1 | 115 | UTR 5 | XP_011526385.1 | |||
XM_011528084.2 | 115 | UTR 5 | XP_011526386.1 |
HOMER3 - homer scaffolding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |