Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAAGAGGGAGGGGCTGGGGGCAC[A/G]GAGGGCCAGTGGCATCTCCTCTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607993 | ||||||||||||||||||||
Literature Links: |
SUGP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SUGP2 - SURP and G-patch domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017392.4 | 5851 | UTR 3 | NP_001017392.2 | |||
NM_001321697.1 | 5851 | Intron | NP_001308626.1 | |||
NM_001321698.1 | 5851 | Intron | NP_001308627.1 | |||
NM_001321699.1 | 5851 | UTR 3 | NP_001308628.1 | |||
NM_014884.4 | 5851 | Intron | NP_055699.2 | |||
XM_017026137.1 | 5851 | UTR 3 | XP_016881626.1 |