Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGCAGGAACCGCGGCTGCTGGA[C/G]AAGAGGGGTGCGGTGGATACTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615956 MIM: 177060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC151 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC151 - coiled-coil domain containing 151 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302453.1 | 21 | Intron | NP_001289382.1 | |||
NM_001302454.1 | 21 | Intron | NP_001289383.1 | |||
NM_145045.4 | 21 | Intron | NP_659482.3 | |||
XM_017026241.1 | 21 | Intron | XP_016881730.1 |
PRKCSH - protein kinase C substrate 80K-H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001329.2 | 21 | UTR 5 | NP_001001329.1 | |||
NM_001289102.1 | 21 | Intron | NP_001276031.1 | |||
NM_001289103.1 | 21 | UTR 5 | NP_001276032.1 | |||
NM_001289104.1 | 21 | UTR 5 | NP_001276033.1 | |||
NM_002743.3 | 21 | UTR 5 | NP_002734.2 | |||
XM_011528130.1 | 21 | UTR 5 | XP_011526432.1 | |||
XM_011528131.1 | 21 | UTR 5 | XP_011526433.1 | |||
XM_011528132.1 | 21 | UTR 5 | XP_011526434.1 | |||
XM_017026977.1 | 21 | UTR 5 | XP_016882466.1 |