Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCGGGGCAAATGTGGAATGCT[C/T]TTTCACTATGACTGGATTAGCGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601913 MIM: 607335 MIM: 607824 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASNA1 - arsA arsenite transporter, ATP-binding, homolog 1 (bacterial) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BEST2 - bestrophin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017682.2 | 872 | Silent Mutation | CTC,CTT | L,L 224 | NP_060152.2 | |
XM_005259963.3 | 872 | Silent Mutation | CTC,CTT | L,L 224 | XP_005260020.1 |
HOOK2 - hook microtubule-tethering protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |