Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAAGCATCCAGAATCCTCTCAC[A/G]GTGGGGTCACACACCCCATGCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608349 MIM: 165040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSH2D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSH2D - hematopoietic SH2 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291274.1 | 155 | UTR 5 | NP_001278203.1 | |||
NM_032855.3 | 155 | UTR 5 | NP_116244.1 | |||
XM_011528386.2 | 155 | Intron | XP_011526688.1 | |||
XM_011528387.2 | 155 | Intron | XP_011526689.1 | |||
XM_011528388.2 | 155 | Intron | XP_011526690.1 | |||
XM_011528389.2 | 155 | Intron | XP_011526691.1 | |||
XM_011528390.1 | 155 | Intron | XP_011526692.1 | |||
XM_017027402.1 | 155 | Intron | XP_016882891.1 | |||
XM_017027403.1 | 155 | Intron | XP_016882892.1 | |||
XM_017027404.1 | 155 | Intron | XP_016882893.1 |
RAB8A - RAB8A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005370.4 | 155 | Intron | NP_005361.2 | |||
XM_011528020.2 | 155 | Intron | XP_011526322.1 |