Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACAAAAAGCAGAAAAGAAAAAG[A/C]AAAGAGTCAAGTGAGCAAAGTGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COLGALT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COLGALT1 - collagen beta(1-O)galactosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024656.3 | Intron | NP_078932.2 | ||||
XM_005260080.4 | Intron | XP_005260137.1 | ||||
XM_011528297.2 | Intron | XP_011526599.1 | ||||
XM_011528298.2 | Intron | XP_011526600.1 | ||||
XM_011528299.2 | Intron | XP_011526601.1 |
FAM129C - family with sequence similarity 129 member C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |