Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCGGCGAGGAGACCCCTGCACC[C/T]TAGGGTGAGTGAATCCCAGACTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNN1 - potassium calcium-activated channel subfamily N member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002248.4 | 225 | UTR 5 | NP_002239.2 | |||
XM_005259905.1 | 225 | Intron | XP_005259962.1 | |||
XM_005259906.1 | 225 | Intron | XP_005259963.1 | |||
XM_005259907.2 | 225 | Intron | XP_005259964.1 | |||
XM_011528004.1 | 225 | Intron | XP_011526306.1 | |||
XM_011528005.2 | 225 | UTR 5 | XP_011526307.1 |