Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCAAATCTTAGAGAACATTATT[A/G]TTCAATGCTTTCATTCATTTACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601604 MIM: 612258 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL12RB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL12RB1 - interleukin 12 receptor subunit beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAST3 - microtubule associated serine/threonine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015016.1 | Intron | NP_055831.1 | ||||
XM_005259823.3 | Intron | XP_005259880.1 | ||||
XM_005259824.2 | Intron | XP_005259881.1 | ||||
XM_005259825.2 | Intron | XP_005259882.1 | ||||
XM_005259826.3 | Intron | XP_005259883.1 | ||||
XM_005259827.2 | Intron | XP_005259884.1 | ||||
XM_006722698.2 | Intron | XP_006722761.1 | ||||
XM_006722699.2 | Intron | XP_006722762.2 | ||||
XM_006722700.3 | Intron | XP_006722763.1 | ||||
XM_011527823.1 | Intron | XP_011526125.1 | ||||
XM_017026506.1 | Intron | XP_016881995.1 | ||||
XM_017026507.1 | Intron | XP_016881996.1 | ||||
XM_017026508.1 | Intron | XP_016881997.1 | ||||
XM_017026509.1 | Intron | XP_016881998.1 | ||||
XM_017026510.1 | Intron | XP_016881999.1 | ||||
XM_017026511.1 | Intron | XP_016882000.1 | ||||
XM_017026512.1 | Intron | XP_016882001.1 | ||||
XM_017026513.1 | Intron | XP_016882002.1 | ||||
XM_017026514.1 | Intron | XP_016882003.1 | ||||
XM_017026515.1 | Intron | XP_016882004.1 | ||||
XM_017026516.1 | Intron | XP_016882005.1 |