Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATCCCGGTTCCAGATTTCCGCG[C/G]TCCAACGCCTCCGCAGCAGCCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604140 MIM: 611625 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIDO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIDO1 - death inducer-obliterator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193369.1 | 105 | Intron | NP_001180298.1 | |||
NM_001193370.1 | 105 | Intron | NP_001180299.1 | |||
NM_022105.4 | 105 | UTR 5 | NP_071388.2 | |||
NM_033081.2 | 105 | UTR 5 | NP_149072.2 | |||
NM_080796.3 | 105 | Intron | NP_542986.1 | |||
NM_080797.3 | 105 | UTR 5 | NP_542987.2 | |||
XM_011528504.1 | 105 | Intron | XP_011526806.1 | |||
XM_011528505.1 | 105 | UTR 5 | XP_011526807.1 | |||
XM_011528506.1 | 105 | Intron | XP_011526808.1 | |||
XM_011528507.1 | 105 | Intron | XP_011526809.1 | |||
XM_011528508.1 | 105 | Intron | XP_011526810.1 | |||
XM_011528509.2 | 105 | Intron | XP_011526811.1 |
GID8 - GID complex subunit 8 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017896.2 | 105 | Intron | NP_060366.1 |