Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTTCTTTTAGAGATTGGTTCTT[G/T]GTGAAAAATTTTTTAGCATCCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASXL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASXL1 - additional sex combs like 1, transcriptional regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164603.1 | Intron | NP_001158075.1 | ||||
NM_015338.5 | Intron | NP_056153.2 | ||||
XM_006723727.3 | Intron | XP_006723790.1 | ||||
XM_006723728.3 | Intron | XP_006723791.1 | ||||
XM_006723730.3 | Intron | XP_006723793.1 | ||||
XM_006723732.2 | Intron | XP_006723795.1 | ||||
XM_006723733.1 | Intron | XP_006723796.1 | ||||
XM_011528648.2 | Intron | XP_011526950.1 | ||||
XM_011528652.2 | Intron | XP_011526954.1 | ||||
XM_017027704.1 | Intron | XP_016883193.1 | ||||
XM_017027705.1 | Intron | XP_016883194.1 | ||||
XM_017027706.1 | Intron | XP_016883195.1 |