Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCAGCCCATTCCAAGCTGCTTA[C/T]TTGAAACTGCATGTAACATGTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605267 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JPH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JPH2 - junctophilin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSER1 - oxidative stress responsive serine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016470.7 | 1770 | UTR 3 | NP_057554.4 | |||
XM_011528851.1 | 1770 | Intron | XP_011527153.1 | |||
XM_011528853.2 | 1770 | Intron | XP_011527155.1 | |||
XM_011528854.2 | 1770 | Intron | XP_011527156.1 | |||
XM_017027873.1 | 1770 | Intron | XP_016883362.1 |