Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCAACAGAGGTGGTTGGCTCGAA[C/G]TCAAACTAAAATGGCCTCAAAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616703 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COMMD7 - COMM domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099339.1 | 1694 | UTR 3 | NP_001092809.1 | |||
NM_053041.2 | 1694 | UTR 3 | NP_444269.2 | |||
XM_005260299.3 | 1694 | Intron | XP_005260356.1 | |||
XM_005260300.3 | 1694 | Intron | XP_005260357.1 | |||
XM_011528604.2 | 1694 | UTR 3 | XP_011526906.1 | |||
XM_017027685.1 | 1694 | UTR 3 | XP_016883174.1 | |||
XM_017027686.1 | 1694 | UTR 3 | XP_016883175.1 | |||
XM_017027687.1 | 1694 | Intron | XP_016883176.1 |