Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCGCCCCATGCTGGGCACTTG[A/G]GGCATGCTGACTGTGGTGAATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606411 MIM: 608679 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC13A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC13A3 - solute carrier family 13 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011554.2 | 2549 | Intron | NP_001011554.1 | |||
NM_001193339.1 | 2549 | Intron | NP_001180268.1 | |||
NM_001193340.1 | 2549 | Intron | NP_001180269.1 | |||
NM_001193342.1 | 2549 | Intron | NP_001180271.1 | |||
NM_022829.5 | 2549 | Intron | NP_073740.2 |
TP53RK - TP53 regulating kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033550.3 | 2549 | UTR 3 | NP_291028.3 |