Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAATTGATGTTTTGGGACCCTTT[C/T]CCATTATCTAATTATGCTTTCTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC9A8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC9A8 - solute carrier family 9 member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260491.1 | Intron | NP_001247420.1 | ||||
NM_015266.2 | Intron | NP_056081.1 | ||||
XM_006723756.1 | Intron | XP_006723819.1 | ||||
XM_011528736.1 | Intron | XP_011527038.1 | ||||
XM_011528737.1 | Intron | XP_011527039.1 | ||||
XM_011528738.1 | Intron | XP_011527040.1 | ||||
XM_011528739.1 | Intron | XP_011527041.1 | ||||
XM_011528740.1 | Intron | XP_011527042.1 | ||||
XM_011528741.1 | Intron | XP_011527043.1 | ||||
XM_011528742.1 | Intron | XP_011527044.1 | ||||
XM_011528743.2 | Intron | XP_011527045.1 | ||||
XM_011528744.2 | Intron | XP_011527046.1 | ||||
XM_011528745.1 | Intron | XP_011527047.1 | ||||
XM_017027754.1 | Intron | XP_016883243.1 | ||||
XM_017027755.1 | Intron | XP_016883244.1 | ||||
XM_017027756.1 | Intron | XP_016883245.1 |