Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTGAGCCCTTGGAGGGGCCTGA[A/G]TGCACCAGTCTGGGGGTTGGGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608200 MIM: 601017 | ||||||||||||||||||||
Literature Links: |
CDK5RAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK5RAP1 - CDK5 regulatory subunit associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNTA1 - syntrophin alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003098.2 | 1283 | Silent Mutation | CAC,CAT | H,H 350 | NP_003089.1 | |
XM_005260517.1 | 1283 | Silent Mutation | CAC,CAT | H,H 350 | XP_005260574.1 | |
XM_011529007.1 | 1283 | Silent Mutation | CAC,CAT | H,H 350 | XP_011527309.1 | |
XM_011529008.1 | 1283 | Silent Mutation | CAC,CAT | H,H 350 | XP_011527310.1 |