Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTAATTTAGGCAAAACCCCCCC[A/G]CCACTGAATCGTCCAACAAAAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927770 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927770 - uncharacterized LOC101927770 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF217 - zinc finger protein 217 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006526.2 | 4433 | UTR 3 | NP_006517.1 | |||
XM_005260545.3 | 4433 | UTR 3 | XP_005260602.1 | |||
XM_006723875.2 | 4433 | UTR 3 | XP_006723938.1 | |||
XM_011529036.1 | 4433 | UTR 3 | XP_011527338.1 | |||
XM_017028059.1 | 4433 | UTR 3 | XP_016883548.1 |