Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATTCAGTGCATTGCTGGAACA[A/G]TTAGAAGTGCTCTGTAACAAAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AURKA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AURKA - aurora kinase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323303.1 | Intron | NP_001310232.1 | ||||
NM_001323304.1 | Intron | NP_001310233.1 | ||||
NM_001323305.1 | Intron | NP_001310234.1 | ||||
NM_003600.3 | Intron | NP_003591.2 | ||||
NM_198433.2 | Intron | NP_940835.1 | ||||
NM_198434.2 | Intron | NP_940836.1 | ||||
NM_198435.2 | Intron | NP_940837.1 | ||||
NM_198436.2 | Intron | NP_940838.1 | ||||
NM_198437.2 | Intron | NP_940839.1 | ||||
XM_017028034.1 | Intron | XP_016883523.1 | ||||
XM_017028035.1 | Intron | XP_016883524.1 |
FAM210B - family with sequence similarity 210 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |