Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGCCTGGGCCGGGGCTGGCTGGG[A/T]TGAGGGGATGGGAGGACCGGGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608725 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHACTR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHACTR3 - phosphatase and actin regulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199505.1 | Intron | NP_001186434.1 | ||||
NM_001199506.1 | Intron | NP_001186435.1 | ||||
NM_001281507.1 | Intron | NP_001268436.1 | ||||
NM_080672.4 | Intron | NP_542403.1 | ||||
NM_183244.1 | Intron | NP_899067.1 | ||||
NM_183246.1 | Intron | NP_899069.1 | ||||
XM_011528525.2 | Intron | XP_011526827.1 | ||||
XM_011528526.2 | Intron | XP_011526828.1 | ||||
XM_017027626.1 | Intron | XP_016883115.1 | ||||
XM_017027627.1 | Intron | XP_016883116.1 | ||||
XM_017027628.1 | Intron | XP_016883117.1 | ||||
XM_017027629.1 | Intron | XP_016883118.1 | ||||
XM_017027630.1 | Intron | XP_016883119.1 | ||||
XM_017027631.1 | Intron | XP_016883120.1 |