Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGTAGGTTCATCCTGAGTCATT[C/T]GGTGAGAGCCCCTGGCGTAGCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSBPL2 - oxysterol binding protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278649.1 | Intron | NP_001265578.1 | ||||
NM_014835.3 | Intron | NP_055650.1 | ||||
NM_144498.2 | Intron | NP_653081.1 | ||||
XM_017028163.1 | Intron | XP_016883652.1 | ||||
XM_017028164.1 | Intron | XP_016883653.1 | ||||
XM_017028165.1 | Intron | XP_016883654.1 | ||||
XM_017028166.1 | Intron | XP_016883655.1 | ||||
XM_017028167.1 | Intron | XP_016883656.1 | ||||
XM_017028168.1 | Intron | XP_016883657.1 | ||||
XM_017028169.1 | Intron | XP_016883658.1 | ||||
XM_017028170.1 | Intron | XP_016883659.1 |