Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGTATTTTAAACACACTGAGCC[A/T]GGGATTACTTGATAGCCATAAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604205 MIM: 607179 | ||||||||||||||||||||
Literature Links: |
CPNE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPNE1 - copine 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198863.1 | 2921 | Intron | NP_001185792.1 | |||
NM_003915.5 | 2921 | Intron | NP_003906.2 | |||
NM_152925.2 | 2921 | Intron | NP_690902.1 | |||
NM_152926.2 | 2921 | Intron | NP_690903.1 | |||
NM_152927.2 | 2921 | Intron | NP_690904.1 | |||
NM_152928.2 | 2921 | Intron | NP_690905.1 |
RBM12 - RNA binding motif protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198838.1 | 2921 | Silent Mutation | CCA,CCT | P,P 883 | NP_001185767.1 | |
NM_001198840.1 | 2921 | Silent Mutation | CCA,CCT | P,P 883 | NP_001185769.1 | |
NM_006047.5 | 2921 | Silent Mutation | CCA,CCT | P,P 883 | NP_006038.2 | |
NM_152838.3 | 2921 | Silent Mutation | CCA,CCT | P,P 883 | NP_690051.1 |