Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAAAGGGAGGCGATACAATGAC[A/G]TGAAACCATAGAGGTAAGAAGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAMHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAMHD1 - SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015474.3 | 2228 | Intron | NP_056289.2 | |||
XM_005260384.3 | 2228 | Intron | XP_005260441.1 | |||
XM_011528761.2 | 2228 | UTR 3 | XP_011527063.1 |
TLDC2 - TBC/LysM-associated domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304783.1 | 2228 | UTR 3 | NP_001291712.1 | |||
NM_080628.2 | 2228 | UTR 3 | NP_542195.1 | |||
XM_017027672.1 | 2228 | Intron | XP_016883161.1 | |||
XM_017027673.1 | 2228 | Intron | XP_016883162.1 | |||
XM_017027674.1 | 2228 | Intron | XP_016883163.1 |