Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAATCTCTTTTGAAGCTGTCCTT[A/G]ACCTGTTTCTCAAGACTGGAATTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116957 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MROH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MROH8 - maestro heat like repeat family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152503.5 | 3433 | UTR 3 | NP_689716.4 | |||
NM_213631.2 | 3433 | Intron | NP_998796.1 | |||
NM_213632.2 | 3433 | Intron | NP_998797.2 |
RBL1 - RB transcriptional corepressor like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |