Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCAAGGCCGTTACGAAACACACA[G/T]GTACTCTCCCACTCACTCAGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613110 MIM: 603106 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLCAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BLCAP - bladder cancer associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167820.1 | 1435 | UTR 3 | NP_001161292.1 | |||
NM_001167821.1 | 1435 | UTR 3 | NP_001161293.1 | |||
NM_001167822.2 | 1435 | UTR 3 | NP_001161294.1 | |||
NM_001167823.1 | 1435 | UTR 3 | NP_001161295.1 | |||
NM_001317074.1 | 1435 | UTR 3 | NP_001304003.1 | |||
NM_001317075.1 | 1435 | UTR 3 | NP_001304004.1 | |||
NM_006698.3 | 1435 | UTR 3 | NP_006689.1 | |||
XM_017027599.1 | 1435 | UTR 3 | XP_016883088.1 |
NNAT - neuronatin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |