Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGGCTCAGTTAACTCCTGCAGT[C/T]AAGTGCTGTCTGGGCAGGCAGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R16B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R16B - protein phosphatase 1 regulatory subunit 16B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172735.2 | Intron | NP_001166206.1 | ||||
NM_015568.3 | Intron | NP_056383.1 | ||||
XM_011528768.2 | Intron | XP_011527070.1 | ||||
XM_011528769.2 | Intron | XP_011527071.1 | ||||
XM_017027785.1 | Intron | XP_016883274.1 |