Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTTGCTCTGCTGATCTCCCAGC[C/T]GTCTCTGGATCAATGGCCTTTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCK - HCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172129.1 | Intron | NP_001165600.1 | ||||
NM_001172130.1 | Intron | NP_001165601.1 | ||||
NM_001172131.1 | Intron | NP_001165602.1 | ||||
NM_001172132.1 | Intron | NP_001165603.1 | ||||
NM_001172133.1 | Intron | NP_001165604.1 | ||||
NM_002110.3 | Intron | NP_002101.2 |