Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAACTCCTGTCAGTTGTATGAAG[A/G]AAATTGCAGTAATATCCACCACTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPTLC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPTLC3 - serine palmitoyltransferase long chain base subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018327.2 | Intron | NP_060797.2 | ||||
XM_011529279.1 | Intron | XP_011527581.1 | ||||
XM_011529280.2 | Intron | XP_011527582.1 | ||||
XM_017027922.1 | Intron | XP_016883411.1 |