Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGAACCTTTGCTAATTCTGGATT[C/T]GAAGGAAAACATTTTAGTTCCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALGAPA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RALGAPA2 - Ral GTPase activating protein catalytic alpha subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020343.3 | Intron | NP_065076.2 | ||||
XM_005260768.2 | Intron | XP_005260825.1 | ||||
XM_006723598.1 | Intron | XP_006723661.1 | ||||
XM_006723599.1 | Intron | XP_006723662.1 | ||||
XM_011529309.1 | Intron | XP_011527611.1 | ||||
XM_011529310.2 | Intron | XP_011527612.1 | ||||
XM_011529311.2 | Intron | XP_011527613.2 | ||||
XM_017027972.1 | Intron | XP_016883461.1 | ||||
XM_017027973.1 | Intron | XP_016883462.1 | ||||
XM_017027974.1 | Intron | XP_016883463.1 | ||||
XM_017027975.1 | Intron | XP_016883464.1 | ||||
XM_017027976.1 | Intron | XP_016883465.1 | ||||
XM_017027977.1 | Intron | XP_016883466.1 |