Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCCCCCAGAGCATGGGCCACA[A/G]CAGTGGTGTCTGCGCATAGTGGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609935 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EBF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EBF4 - early B-cell factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110514.1 | Intron | NP_001103984.1 | ||||
XM_006723600.1 | Intron | XP_006723663.1 | ||||
XM_017027983.1 | Intron | XP_016883472.1 | ||||
XM_017027984.1 | Intron | XP_016883473.1 | ||||
XM_017027985.1 | Intron | XP_016883474.1 |