Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGTCAAACACACCCGTGGTCAC[A/G]TCATGGTTAATCCCATCCACGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPXM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPXM1 - carboxypeptidase X (M14 family), member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184699.1 | 1805 | Silent Mutation | GAC,GAT | D,D 577 | NP_001171628.1 | |
NM_019609.4 | 1805 | Silent Mutation | GAC,GAT | D,D 651 | NP_062555.1 |