Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAAATATCAGTGGCTGTGACTCC[C/T]CTGTCCCCCACACCAATTATGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614146 MIM: 610206 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf194 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C20orf194 - chromosome 20 open reading frame 194 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009984.2 | 4769 | UTR 3 | NP_001009984.1 | |||
XM_005260684.3 | 4769 | UTR 3 | XP_005260741.1 | |||
XM_005260687.4 | 4769 | UTR 3 | XP_005260744.1 | |||
XM_011529208.2 | 4769 | UTR 3 | XP_011527510.1 | |||
XM_017027783.1 | 4769 | UTR 3 | XP_016883272.1 |
SLC4A11 - solute carrier family 4 member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |