Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCTCACGCTCTCACCCATGAC[A/G]CGGGCCTGCACCGTCACGCGCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610702 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C20orf27 - chromosome 20 open reading frame 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039140.2 | 1100 | Silent Mutation | CGC,CGT | R,R 164 | NP_001034229.1 | |
NM_001258429.1 | 1100 | Silent Mutation | CGC,CGT | R,R 139 | NP_001245358.1 | |
NM_001258430.1 | 1100 | Silent Mutation | CGC,CGT | R,R 139 | NP_001245359.1 | |
XM_011529266.2 | 1100 | Intron | XP_011527568.1 |
HSPA12B - heat shock protein family A (Hsp70) member 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |