Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGTAATGCAGAAATAGATAACC[A/G]GAAGAGTGACTAAGCCAGGAAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM230 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM230 - transmembrane protein 230 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009923.1 | Intron | NP_001009923.1 | ||||
NM_001009924.1 | Intron | NP_001009924.1 | ||||
NM_001009925.1 | Intron | NP_001009925.1 | ||||
NM_014145.4 | Intron | NP_054864.3 | ||||
XM_005260713.3 | Intron | XP_005260770.1 | ||||
XM_011529227.2 | Intron | XP_011527529.1 | ||||
XM_011529229.2 | Intron | XP_011527531.1 | ||||
XM_017027834.1 | Intron | XP_016883323.1 | ||||
XM_017027835.1 | Intron | XP_016883324.1 | ||||
XM_017027836.1 | Intron | XP_016883325.1 |