Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGGGTTGGAGTGGTGGGGGCGG[G/T]GGTCACAGCAATGCATCTGATCGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf196 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C20orf196 - chromosome 20 open reading frame 196 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303477.1 | Intron | NP_001290406.1 | ||||
NM_001303478.1 | Intron | NP_001290407.1 | ||||
NM_001303479.1 | Intron | NP_001290408.1 | ||||
NM_152504.3 | Intron | NP_689717.2 | ||||
XM_006723549.3 | Intron | XP_006723612.1 | ||||
XM_011529177.2 | Intron | XP_011527479.1 | ||||
XM_011529178.2 | Intron | XP_011527480.1 | ||||
XM_011529179.2 | Intron | XP_011527481.1 | ||||
XM_011529180.2 | Intron | XP_011527482.1 | ||||
XM_011529181.2 | Intron | XP_011527483.1 | ||||
XM_011529182.2 | Intron | XP_011527484.1 | ||||
XM_017027684.1 | Intron | XP_016883173.1 |