Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGTAAACTGTAAATGTTTCCTT[A/C]TCAGAATTGTTTCATGTTATCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLCB1 - phospholipase C beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015192.3 | Intron | NP_056007.1 | ||||
NM_182734.2 | Intron | NP_877398.1 | ||||
XM_011529199.2 | Intron | XP_011527501.1 | ||||
XM_011529200.2 | Intron | XP_011527502.1 | ||||
XM_011529201.2 | Intron | XP_011527503.1 | ||||
XM_011529203.2 | Intron | XP_011527505.1 | ||||
XM_017027752.1 | Intron | XP_016883241.1 | ||||
XM_017027753.1 | Intron | XP_016883242.1 |