Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCATTCACACCAGCACAATGGTG[C/T]TCCTCCAAGAATGTATTTATTTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104760 | ||||||||||||||||||||
Literature Links: |
APP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APP - amyloid beta precursor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000484.3 | 3520 | UTR 3 | NP_000475.1 | |||
NM_001136016.3 | 3520 | UTR 3 | NP_001129488.1 | |||
NM_001136129.2 | 3520 | UTR 3 | NP_001129601.1 | |||
NM_001136130.2 | 3520 | UTR 3 | NP_001129602.1 | |||
NM_001136131.2 | 3520 | UTR 3 | NP_001129603.1 | |||
NM_001204301.1 | 3520 | UTR 3 | NP_001191230.1 | |||
NM_001204302.1 | 3520 | UTR 3 | NP_001191231.1 | |||
NM_001204303.1 | 3520 | UTR 3 | NP_001191232.1 | |||
NM_201413.2 | 3520 | UTR 3 | NP_958816.1 | |||
NM_201414.2 | 3520 | UTR 3 | NP_958817.1 |