Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGCATATTCATTTAGCATTCAC[A/G]TTAAACCCACATTTGACTCTAACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAXBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAXBP1 - PAX3 and PAX7 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013329.3 | 3557 | Intron | NP_037461.2 | |||
NM_016631.3 | 3557 | UTR 3 | NP_057715.2 | |||
XM_006724066.1 | 3557 | UTR 3 | XP_006724129.1 | |||
XM_011529804.2 | 3557 | Intron | XP_011528106.1 |
PAXBP1-AS1 - PAXBP1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNJ1 - synaptojanin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |