Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCAGCAGGGAGAGACCCGGACA[C/T]CTGTCATCGCTTCTCGGTGGATCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603796 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNE2 - potassium voltage-gated channel, Isk-related family, member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMIM11 - small integral membrane protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_058182.4 | Intron | NP_478062.1 |