Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCCTGTGAAATATTCATGTGC[A/G]TCACCCACTTTGCCTTAGGTTCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176261 MIM: 602917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNE1 - potassium voltage-gated channel subfamily E regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000219.5 | Intron | NP_000210.2 | ||||
NM_001127668.3 | Intron | NP_001121140.1 | ||||
NM_001127669.3 | Intron | NP_001121141.1 | ||||
NM_001127670.3 | Intron | NP_001121142.1 | ||||
NM_001270402.2 | Intron | NP_001257331.1 | ||||
NM_001270403.2 | Intron | NP_001257332.1 | ||||
NM_001270404.2 | Intron | NP_001257333.1 | ||||
NM_001270405.2 | Intron | NP_001257334.1 | ||||
XM_017028342.1 | Intron | XP_016883831.1 |
RCAN1 - regulator of calcineurin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |