Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGTTCAAATAGGAAAAAAAAAA[C/G]TGTGTTATTTTCTTAAGACAAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147150 MIM: 602060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MX1 - MX dynamin like GTPase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS2 - transmembrane protease, serine 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135099.1 | Intron | NP_001128571.1 | ||||
NM_005656.3 | Intron | NP_005647.3 | ||||
XM_005261043.2 | Intron | XP_005261100.1 | ||||
XM_011529731.2 | Intron | XP_011528033.1 | ||||
XM_011529732.2 | Intron | XP_011528034.1 | ||||
XM_011529733.1 | Intron | XP_011528035.1 |