Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCAGTCACCGAACTGGGTTTC[A/G]CTTCCATTTCCTTACTTGTTTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605674 MIM: 602621 | ||||||||||||||||||||
Literature Links: |
BTG3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTG3 - BTG anti-proliferation factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130914.1 | 831 | Missense Mutation | NP_001124386.1 | |||
NM_006806.4 | 831 | Missense Mutation | NP_006797.3 | |||
XM_011529441.1 | 831 | Missense Mutation | XP_011527743.1 | |||
XM_017028265.1 | 831 | Missense Mutation | XP_016883754.1 | |||
XM_017028266.1 | 831 | Missense Mutation | XP_016883755.1 |
CXADR - coxsackie virus and adenovirus receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207063.1 | 831 | Intron | NP_001193992.1 | |||
NM_001207064.1 | 831 | Intron | NP_001193993.1 | |||
NM_001207065.1 | 831 | Intron | NP_001193994.1 | |||
NM_001207066.1 | 831 | Intron | NP_001193995.1 | |||
NM_001338.4 | 831 | Intron | NP_001329.1 | |||
XM_011529476.2 | 831 | Intron | XP_011527778.1 | |||
XM_011529477.2 | 831 | Intron | XP_011527779.1 | |||
XM_011529478.2 | 831 | Intron | XP_011527780.1 | |||
XM_011529479.1 | 831 | Intron | XP_011527781.1 |