Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTTACTACACAGACTCAGTAG[C/T]GGAAAAACATATCCCATAAGCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605924 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR4 - WD repeat domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260474.1 | 1145 | Intron | NP_001247403.1 | |||
NM_001260475.1 | 1145 | Intron | NP_001247404.1 | |||
NM_001260476.1 | 1145 | Intron | NP_001247405.1 | |||
NM_001260477.1 | 1145 | Intron | NP_001247406.1 | |||
NM_018669.5 | 1145 | Intron | NP_061139.2 | |||
NM_033661.4 | 1145 | UTR 3 | NP_387510.1 | |||
XM_011529433.1 | 1145 | Intron | XP_011527735.1 | |||
XM_017028262.1 | 1145 | Intron | XP_016883751.1 | |||
XM_017028263.1 | 1145 | Intron | XP_016883752.1 | |||
XM_017028264.1 | 1145 | UTR 3 | XP_016883753.1 |