Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCTTATTCCTTCTCCAGAGCA[A/C]CTGTTCCTTCAGGCCTTCTGGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610653 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RRP1 - ribosomal RNA processing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003683.5 | 363 | Missense Mutation | CAA,CAC | Q,H 93 | NP_003674.1 | |
XM_017028485.1 | 363 | Missense Mutation | CAA,CAC | Q,H 93 | XP_016883974.1 | |
XM_017028486.1 | 363 | UTR 5 | XP_016883975.1 |