Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCAGCAGCAGGAGGTCATCTC[A/G]CCGGACACTACCGAGAGAAACAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL6A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL6A2 - collagen type VI alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001849.3 | 454 | Silent Mutation | TCA,TCG | S,S 27 | NP_001840.3 | |
NM_058174.2 | 454 | Silent Mutation | TCA,TCG | S,S 27 | NP_478054.2 | |
NM_058175.2 | 454 | Silent Mutation | TCA,TCG | S,S 27 | NP_478055.2 | |
XM_011529451.1 | 454 | Silent Mutation | TCA,TCG | S,S 27 | XP_011527753.1 |