Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACCTGGAACCCTGCTTAATGGT[C/G]GTGAAGACGTTGATGGATGTCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613859 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UMODL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UMODL1 - uromodulin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004416.2 | Intron | NP_001004416.2 | ||||
NM_001199527.1 | Intron | NP_001186456.1 | ||||
NM_001199528.2 | Intron | NP_001186457.2 | ||||
NM_173568.3 | Intron | NP_775839.3 | ||||
XM_011529797.1 | Intron | XP_011528099.1 | ||||
XM_017028506.1 | Intron | XP_016883995.1 | ||||
XM_017028507.1 | Intron | XP_016883996.1 | ||||
XM_017028508.1 | Intron | XP_016883997.1 |
UMODL1-AS1 - UMODL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |